Imidazo[1,2-b]pyridazine ≥98.0%
Leverantör: TCI EUROPE
I0742-1G
I0742-5G
TCIAI0742-1GEA
335
SEK
TCIAI0742-1G
TCIAI0742-5G
Imidazo[1,2-b]pyridazine ≥98.0%
Imidazo[1,2-b]pyridazine
Formel:
C₆H₅N₃ MW: 119,12 g/mol |
MDL Number:
MFCD07782103 CAS nummer: 766-55-2 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...