1,1'-Bis(di-tert-butylphosphino)ferrocene ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1,1'-Bis(di-t-butylphosphino)ferrocene, 1,1'-Bis[bis(1,1-dimethylethyl)phosphino]ferrocene, DTBPF
L19759.04 L19759.ME
L19759.04EA 4525 SEK
L19759.04 L19759.ME
1,1'-Bis(di-tert-butylphosphino)ferrocene ≥98%
1,1'-Bis(di-tert-butylphosphino)ferrocene
Purity: 98% CAS No.: 84680-95-5 Molecular Formula: C26H44FeP2 Formula Weight: 474.43 Melting Point: 73-78° MDL No.: MFCD01630818
Formel: C₂₆H₄₄FeP₂
MW: 474.42 g/mol
Smältpunkt: 73…78 °C
Storage Temperature: Omgivande
MDL Number: MFCD01630818
CAS nummer: 84680-95-5