4-Bromo-1H-pyrazole ≥98.0%
Leverantör: TCI EUROPE
Varning
B3745-5G B3745-25G
TCIAB3745-5GEA 615 SEK
TCIAB3745-5G TCIAB3745-25G
4-Bromo-1H-pyrazole ≥98.0%
4-Bromo-1H-pyrazole
Formel: C₃H₃BrN₂
MW: 146,97 g/mol
Kokpunkt: 260°C
MDL Number: MFCD00075602
CAS nummer: 2075-45-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ff81b418482d7c3c6241a4a13dc3bf410c89fbd&vid=9ff81b418482d7c3c6241a4a13dc3bf410c89fbd&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 95.0 to 99.0 deg-C