Cyclohexylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI EUROPE
Varning

C2188-1G C2188-5G
TCIAC2188-1GEA 335 SEK
TCIAC2188-1G TCIAC2188-5G
Cyclohexylboronic acid (contains varying amounts of Anhydride)
Cyclohexylboronic acid
Formel: C₆H₁₃BO₂
MW: 127,9772 g/mol
Smältpunkt: 120°C
MDL Number: MFCD01311824
CAS nummer: 4441-56-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ef1cc819adb3f96546fe4a6b3eb0709358f23a2&vid=6ef1cc819adb3f96546fe4a6b3eb0709358f23a2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) 97.0 to 116.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR