Ethyl vanillate ≥97%

Leverantör: Thermo Scientific

Synonyms: Ethyl 4-hydroxy-3-methoxybenzoate

L05798.09 L05798.18
L05798.09EA 1450 SEK
L05798.09 L05798.18
Ethyl vanillate ≥97%
Ethyl vanillate

Purity: 97% CAS No.: 617-05-0 Molecular Formula: C10H12O4 Formula Weight: 196.20 Melting Point: 42-46° Boiling Point: 293° MDL No.: MFCD00017269

Formel: C₁₀H₁₂O₄
MW: 196,20 g/mol
Kokpunkt: 293 °C
Smältpunkt: 42…46 °C
Storage Temperature: Omgivande
MDL Number: MFCD00017269
CAS nummer: 617-05-0
EINECS: 210-503-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR