Kaliumhydroxid 0.1 mol/l (0.1 N) isopropanol, Titripur® Volumetric acc. to DIN 51558 part 1, Supelco®
Leverantör: Merck
Fara
Synonyms: Potash caustic, Caustic potash
MQ100
1.05544.1000 1.05544.1003
1.05544.1000EA 527.63 SEK
1.05544.1000 1.05544.1003
Kaliumhydroxid 0.1 mol/l (0.1 N) isopropanol, Titripur® Volumetric acc. to DIN 51558 part 1, Supelco®
Kaliumhydroxid
Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.
Formel: KOH
MW: 56.11 g/mol
Kokpunkt: 1320
Smältpunkt: 360
Storage Temperature: Omgivande
MDL Number: MFCD00003553
CAS nummer: 1310-58-3
EINECS: 215-181-3
UN: 2924
ADR: 3 (8) II
Merck Index: 12,07806