Magnetic stirring bar retrievers, PTFE
Leverantör: SciLabware
SWN564 SWN566
AZLOSWN564EA 475 SEK
AZLOSWN564 AZLOSWN566
Magnetic stirring bar retrievers, PTFE
Omrörare Omrörningsverktyg
PTFE covered. For the removal of stirring magnets from vessels of all kinds.
  • Very good chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +260 °C)