Chiral columns, ReproSil® Chiral-AM
Leverantör: DR MAISCH
R60.AM.S0546 R63.AM.S0546 R65.AM.V0104 R63.AM.S1046 R63.AM.S1546 R60.AM.S2546 R60.AM.S2510 R65.AM.S2510 R65.AM.S1546 R65.AM.S1046 R65.AM.S0546
554-2590EA 0 SEK
554-2590 554-2605 554-2622 554-2606 554-2607 554-2592 554-2591 554-2621 554-2620 554-2619 554-2618
Chiral columns, ReproSil® Chiral-AM
Kromatografikolonn
USP L51 column: amylose tris-3,5-dimethylphenylcarbamate coated on porous silica particles.