Chiral columns, ReproSil® Chiral-OM
Leverantör: DR MAISCH
R65.OM.S2546 R65.OM.S2520 R65.OM.S2510 R65.OM.S1546 R65.OM.S0546 R60.OM.S0546 R60.OM.S2546 R60.OM.S2520 R60.OM.S2510 R60.OM.S1046 R60.OM.S1546 R63.OM.V0104 R63.OM.S1546 R63.OM.S1046
554-2631EA 15725 SEK
554-2631 554-2630 554-2629 554-2628 554-2627 554-2596 554-2601 554-2600 554-2599 554-2597 554-2598 554-2614 554-2613 554-2612
Chiral columns, ReproSil® Chiral-OM
Kromatografikolonn
USP L40 column: cellulose tris-3,5-dimethylphenylcarbamate coated on porous silica particles.