General purpose gloves, Jersette 307
Leverantör: MAPA
34307016 34307017 34307018 34307019
MAPA34307016EA 130 SEK
MAPA34307016 MAPA34307017 MAPA34307018 MAPA34307019
General purpose gloves, Jersette 307
Handskar
Liquidproof natural latex gloves with knitted textile interior for increased comfort.
  • Exceptional comfort, due to the precision of the textile support and the anatomical shape
  • Excellent dexterity and good grip
  • Pebbled exterior finish
  • Provides heat resistance
Ideal for small general maintenance jobs and cleaning tasks involving household detergents.
Certifikat: EN 388:2016 (2120X), EN 407 (X1XXXX); Cat. II