Magnetic hotplate stirrer, hotMIX 1
Leverantör: 2MAG
61001_CH 61001_UK
TMAG61001_CHEA 6225 SEK
TMAG61001_CH TMAG61001_UK
Magnetic hotplate stirrer, hotMIX 1
Omrörare Magnetomrörare med värme
Compact, powerful heatable single point stirrer suitable for the operation with viscous and aggressive media in chemical, biotechnical and medical applications.
  • Hot plate made from resistant aluminium alloy
  • Price-optimised magnetic stirrer
  • 100% maintenance-free and wear-free
  • Speed range from 100 to 1600 min⁻¹
  • Jerk-free stirring even at low speeds
SoftStart for reliable catching/centering and safe accelerating of the stirring bar, resistant stainless steel housing for easy and fast cleaning, spill over protection, extremely robust construction. Stirrer contains three integrated over temperature protection loops to resist against overheating.
Certifikat: Complies with IP 20 protection class.