Methyl-o-toluate

Leverantör: Thermo Scientific

Synonyms: Methyl 2-methylbenzoate, o-Toluic acid methyl ester, 2-Toluic acid methyl ester, Methyl-2-methylbenzoate, 2-Methylbenzoic acid methyl ester

TL00700EA
MAYBTL00700EAEA 0 SEK
MAYBTL00700EA
Methyl-o-toluate
Methyl-o-toluate
Formel: C₉H₁₀O₂
MW: 150,18 g/mol
Kokpunkt: 215 °C (1013 hPa)
Smältpunkt: –50 °C
Densitet: 1,079 g/cm³ (0 °C)
MDL Number: MFCD00008428
CAS nummer: 89-71-4
EINECS: 201-932-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR