2-Brom-4'-pentyloacetophenone
Leverantör: Thermo Scientific
Synonyms: 4-(Pent-1-yl)phenacyl bromide
BTB12841DA BTB12841EA
MAYBBTB12841DAEA 0 SEK
MAYBBTB12841DA MAYBBTB12841EA
2-Brom-4'-pentyloacetophenone
2-Brom-4'-pentyloacetophenone
Formel: C₁₃H₁₇BrO
CAS nummer: 64328-68-3