Ethyl-4-isothiocyanatobenzoate ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: 4-(Ethoxycarbonyl)phenyl isothiocyanate
L12522.09 L12522.04
L12522.09EA 3075 SEK
L12522.09 L12522.04
Ethyl-4-isothiocyanatobenzoate ≥97%
Ethyl-4-isothiocyanatobenzoate
Purity: 97% CAS No.: 1205-06-7 Molecular Formula: C10H9NO2S Formula Weight: 207.25 Odour: Irritating Melting Point: 52-56° Boiling Point: 123-127°/1mm Beilstein Reference: 2213400 UN No.: 2923 MDL No.: MFCD00022058
Formel: C₁₀H₉NO₂S
MW: 207,25 g/mol
Kokpunkt: 123…127 °C (1 mmHg)
Smältpunkt: 52…56 °C
Storage Temperature: Omgivande
MDL Number: MFCD00022058
CAS nummer: 1205-06-7
EINECS: 214-880-0
ADR: 8,III