4-(Benzyloxy)phenyl isothiocyanate
Leverantör: Thermo Scientific
SB01753DA
SB01753EA
MAYBSB01753DAEA
0
SEK
MAYBSB01753DA
MAYBSB01753EA
4-(Benzyloxy)phenyl isothiocyanate
4-(Benzyloxy)phenyl isothiocyanate
Formel:
C₁₄H₁₁NOS MW: 241,31 g/mol |
MDL Number:
MFCD00041095 CAS nummer: 139768-71-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...