4-Fluorostyrene ≥98.0% stabilised
Leverantör: TCI
|
Varning
|
Synonyms:
1-Fluoro-4-vinylbenzene
Stabilization:
med 4-tert-Butylpyrokatekol
F0461-25G
F0461-5G
TCIAF0461-25GEA
2475
SEK
TCIAF0461-25G
TCIAF0461-5G
4-Fluorostyrene ≥98.0% stabilised
4-Fluorostyrene
Formel:
C₈H₇F Kokpunkt: 145 °C (1013 hPa) Densitet: 1,022 g/cm³ (20 °C) Flampunkt: 35°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00000361 CAS nummer: 405-99-2 EINECS: 206-975-0 UN: 1993 ADR: 3,III |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...