(4-Fluorophenyl)methanesulphonyl chloride ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: 4-Fluoro-α-toluenesulphonyl chloride
H31605.MD H31605.03
H31605.MDEA 1200 SEK
H31605.MD H31605.03
(4-Fluorophenyl)methanesulphonyl chloride ≥97%
(4-Fluorophenyl)methanesulphonyl chloride
Purity: 97% CAS No.: 103360-04-9 Molecular Formula: C7H6ClFO2S Formula Weight: 208.64 Melting Point: 58-61° UN No.: 3261 MDL No.: MFCD01631930
Formel: C₇H₆ClFO₂S
MW: 208,64 g/mol
Smältpunkt: 58…61 °C
Storage Temperature: Omgivande
MDL Number: MFCD01631930
CAS nummer: 103360-04-9
ADR: 8,II