2-Butylimidazole ≥97.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

B3136-25G B3136-5G
TCIAB3136-25GEA 2825 SEK
TCIAB3136-25G TCIAB3136-5G
2-Butylimidazole ≥97.0% (by GC, titration analysis)
2-Butylimidazole
Formel: C₇H₁₂N₂
MW: 124,1800 g/mol
Kokpunkt: 294°C
MDL Number: MFCD00792664
CAS nummer: 50790-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fe6aded64a2ccda4eda4e66b395186a529ffe4e&vid=1fe6aded64a2ccda4eda4e66b395186a529ffe4e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 51.0 to 56.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR