2-Methyl-2-imidazoline ≥97.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 4,5-Dihydro-2-methyl-1H-imidazole, 2-Methyl-4,5-dihydro-1H-imidazole, Methylglyoxalidine, 2-Methylimidazoline, Lysidine

M0579-25G M0579-5G
TCIAM0579-25GEA 4425 SEK
TCIAM0579-25G TCIAM0579-5G
2-Methyl-2-imidazoline ≥97.0% (by GC, titration analysis)
2-Methyl-2-imidazoline
Formel: C₄H₈N₂
MW: 84,12 g/mol
Kokpunkt: 130°C
MDL Number: MFCD00051490
CAS nummer: 534-26-9
EINECS: 208-596-6
Merck Index: 14,05635
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a93caefa3ff4d4834a31f6a43f17f8b1b92a2655&vid=83169f5e29590102add8034a37631de40d605669&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 102.0 to 105.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR