1H-Pyrazole-3-boronic acid pinacol ester 95%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 2-(1H-Pyrazol-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1H-Pyrazole-3-boronic acid Pinacol ester
H27619.06 H27619.03
H27619.06EA 20300 SEK
H27619.06 H27619.03
1H-Pyrazole-3-boronic acid pinacol ester 95%
1H-Pyrazole-3-boronic acid pinacol ester
Purity: 95% CAS No.: 844501-71-9 Molecular Formula: C9H15BN2O2 Formula Weight: 194.04 Melting Point: 105-108° MDL No.: MFCD07368044
Formel: C₉H₁₅BN₂O₂
MW: 194.04 g/mol
Smältpunkt: 105…108 °C
Storage Temperature: Omgivande
MDL Number: MFCD07368044
CAS nummer: 844501-71-9