2-Butylimidazole-4-carbaldehyde

Leverantör: Thermo Scientific

Synonyms: 2-Butyl-1H-imidazole-5-carboxaldehyde

SB02028DA SB02028EA SB02028EE
MAYBSB02028DAEA 0 SEK
MAYBSB02028DA MAYBSB02028EA MAYBSB02028EE
2-Butylimidazole-4-carbaldehyde
2-Butylimidazole-4-carbaldehyde
Formel: C₈H₁₂N₂O
MW: 152,193 g/mol
MDL Number: MFCD01075030
CAS nummer: 68282-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR