2-Aminoimidazole hemisulphate 98+%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Aminoimidazole sulphate, 1H-imidazol-2-amine sulphate(2:1)
368860025 368860100
ACRO368860025EA 1400 SEK
ACRO368860025 ACRO368860100
2-Aminoimidazole hemisulphate 98+%
2-Aminoimidazole hemisulphate
Formel: C₆H₁₂N₆O₄S
MW: 264,26 g/mol
Smältpunkt: 270 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00013162
CAS nummer: 1450-93-7
EINECS: 215-918-9
Order Now
Specifikation testresultat
Appearance Brown Crystalline powder or crystals
Infrared spectrum Conforms
Titration with NaOH ≥98.0 %