2-Methylimidazo[4,5-c]pyridine ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Methyl-3H-imidazo[4,5-c]pyridine
L19131.03 L19131.06 L19131.MD
L19131.03EA 2600 SEK
L19131.03 L19131.06 L19131.MD
2-Methylimidazo[4,5-c]pyridine ≥99%
2-Methylimidazo[4,5-c]pyridine
Purity: 99% CAS No.: 63604-59-1 Molecular Formula: C7H7N3 Formula Weight: 133.15 Melting Point: 176-178° Beilstein Reference: 3234
Formel: C₇H₇N₃
MW: 133,15 g/mol
Smältpunkt: 176…178 °C
Storage Temperature: Omgivande
MDL Number: MFCD03426025
CAS nummer: 63604-59-1