(Chloromethylene)dimethylammonium chloride 96%
Leverantör: Thermo Scientific
Fara
Synonyms: N-Chloromethylen-N,N-Dimethyl Ammonium Chloride
295770250 295770050
ACRO295770250EA 2275 SEK
ACRO295770250 ACRO295770050
(Chloromethylene)dimethylammonium chloride 96%
(Chloromethylene)dimethylammonium chloride
Formel: C₃H₇Cl₂N
MW: 128,00 g/mol
Smältpunkt: 139 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00011868
CAS nummer: 3724-43-4
EINECS: 425-970-6
UN: 3261
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Light yellow to light brown Crystalline powder
Infrared spectrum Conforms
Titration Argentometric 95 to 105 %