4-(1-Naphthyl)phenylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI
Varning

Synonyms: 4-(1-Naphthyl)benzeneboronic acid

N0798-1G N0798-5G
TCIAN0798-1GEA 445 SEK
TCIAN0798-1G TCIAN0798-5G
4-(1-Naphthyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(1-Naphthyl)phenylboronic acid
Formel: C₁₆H₁₃BO₂
MW: 248.09 g/mol
MDL Number: MFCD08669639
CAS nummer: 870774-25-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7933fb0b85c159d95e7a64dc243b0542d445ffe2&vid=7933fb0b85c159d95e7a64dc243b0542d445ffe2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 108.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR