Etylakrylat ≥99.0% stabilised
Leverantör: TCI EUROPE
Fara
Synonyms: Acrylic acid-ethyl ester
Stabilization: Stabilised with 4-methoxyphenol (MEHQ)
A0143-25ML A0143-500ML
TCIAA0143-25MLEA 215 SEK
TCIAA0143-25ML TCIAA0143-500ML
Etylakrylat ≥99.0% stabilised
Etylakrylat
Formel: C₅H₈O₂
MW: 100,12 g/mol
Kokpunkt: 100°C
Flampunkt: 8°C
Storage Temperature: Kylskåp
MDL Number: MFCD00009188
CAS nummer: 140-88-5
EINECS: 205-438-8
UN: 1917
ADR: 3,II
Merck Index: 14,03759
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c8b7effa801d2ca885ddbf447ff282a2c0db259&vid=7c8b7effa801d2ca885ddbf447ff282a2c0db259&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless clear liquid
Purity (GC) min. 99.0 %