Natriumoktan-1-sulfonat, vattenfri 99+% för jonparkromatografi
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Sodium octane-1-sulphonate
396691000 396690250
ACRO396691000EA 17975 SEK
ACRO396691000 ACRO396690250
Natriumoktan-1-sulfonat, vattenfri 99+% för jonparkromatografi
Natriumoktan-1-sulfonat
Formel: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007544
CAS nummer: 5324-84-5
EINECS: 226-195-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1244cb11db07775dec2e193823e40080024b7036&vid=32d95cd9e321818e2f94c8261c4b4e6180ee0fc7&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤2 % (150°C)
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.4 M soln.) Passes test
Suitability (HPLC gradient) Passes test