D (-)-arabinos ≥99.0%
Leverantör: TCI EUROPE
Synonyms:
D-(-)-Arabinose, (2S,3R,4R)-2,3,4,5-Tetrahydroxypentanal
A0513-25G
A0513-100G
TCIAA0513-25GEA
495
SEK
TCIAA0513-25G
TCIAA0513-100G
D (-)-arabinos ≥99.0%
D (-)-arabinos
Formel:
C₅H₁₀O₅ MW: 150,13 g/mol Smältpunkt: 160 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00135608 CAS nummer: 10323-20-3 EINECS: 233-708-5 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Melting point | 156.0 to 165.0 deg-C |
Specific rotation [a]20/D | -102 to -106 deg (C=10, H2O) |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...