1-Bromo-2-chloro-5-nitrobenzene ≥97.0%

Leverantör: TCI EUROPE

Synonyms: 3-Bromo-4-chloronitrobenzene

B1624-25G B1624-5G
TCIAB1624-25GEA 2225 SEK
TCIAB1624-25G TCIAB1624-5G
1-Bromo-2-chloro-5-nitrobenzene ≥97.0%
1-Bromo-2-chloro-5-nitrobenzene
Formel: C₆H₃BrClNO₂
MW: 236,4500 g/mol
MDL Number: MFCD00100437
CAS nummer: 16588-26-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a62a7806ebca8553a01d14fd291713e1e1d08591&vid=a62a7806ebca8553a01d14fd291713e1e1d08591&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 57.0 to 60.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR