cis-3-Hexenyl acetate ≥99%
Leverantör: Thermo Scientific
Varning
A19380.14 A19380.22
A19380.14EA 1100 SEK
A19380.14 A19380.22
cis-3-Hexenyl acetate ≥99%
cis-3-Hexenyl acetate
Purity: 99% CAS No.: 3681-71-8 Molecular Formula: C8H14O2 Formula Weight: 142.20 Flash Point: 59°(138°F) Refractive Index: 1.4270 Boiling Point: 169° Beilstein Reference: 1721854 UN No.: 3272 MDL No.: MFCD00036563
Formel: C₈H₁₄O₂
MW: 142,20 g/mol
Kokpunkt: 169 °C
Densitet: 0,899
Flampunkt: 59 °C (138 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00036563
CAS nummer: 3681-71-8
EINECS: 222-960-1
UN: 3272
ADR: 3,III