5-Hexenyl acetate ≥98.0%

Leverantör: TCI EUROPE
Varning

A1338-25ML A1338-500ML
TCIAA1338-25MLEA 590 SEK
TCIAA1338-25ML TCIAA1338-500ML
5-Hexenyl acetate ≥98.0%
5-Hexenyl acetate
Formel: C₈H₁₄O₂
MW: 142,20 g/mol
Kokpunkt: 172°C
MDL Number: MFCD00060856
CAS nummer: 5048-26-0
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3d6405ce5f9828a400607ba2794aacd7bd73afb&vid=c3d6405ce5f9828a400607ba2794aacd7bd73afb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR