(S)-4-Benzyloxy-1,2-butanediol ≥95.0%

Leverantör: TCI

B2900-1G B2900-200MG
TCIAB2900-1GEA 3425 SEK
TCIAB2900-1G TCIAB2900-200MG
(S)-4-Benzyloxy-1,2-butanediol ≥95.0%
(S)-4-Benzyloxy-1,2-butanediol
Formel: C₁₁H₁₆O₃
Kokpunkt: 170°C
MDL Number: MFCD10000950
CAS nummer: 69985-32-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f49c7269b0dfa666eab2d1d1a67bca07140a762&vid=5f49c7269b0dfa666eab2d1d1a67bca07140a762&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %
Specific rotation [a]20/D -20.0 to -25.0 deg (C=5, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR