1-Chloro-2,3-dimethoxybenzene ≥97.0%

Leverantör: TCI EUROPE

C1355-1G C1355-5G
TCIAC1355-1GEA 1300 SEK
TCIAC1355-1G TCIAC1355-5G
1-Chloro-2,3-dimethoxybenzene ≥97.0%
1-Chloro-2,3-dimethoxybenzene
Formel: C₈H₉ClO₂
Kokpunkt: 98°C
MDL Number: MFCD00191399
CAS nummer: 90282-99-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72c8deafaf1cb990a2b9c872e4cb1e73d11af2bd&vid=72c8deafaf1cb990a2b9c872e4cb1e73d11af2bd&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR