2-Amino-6-methoxy-3-nitropyridine ≥97.0%

Leverantör: TCI
Varning

A2372-25G A2372-5G
TCIAA2372-25GEA 4300 SEK
TCIAA2372-25G TCIAA2372-5G
2-Amino-6-methoxy-3-nitropyridine ≥97.0%
2-Amino-6-methoxy-3-nitropyridine
Formel: C₆H₇N₃O₃
MDL Number: MFCD03206481
CAS nummer: 73896-36-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33600a209e9b62471da76353b197066574722a47&vid=33600a209e9b62471da76353b197066574722a47&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 169.0 to 173.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR