2-Hydroxy-5-methylisophthalaldehyde ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Hydroxy-5-methylbenzene-1,3-dicarbaldehyde

H0683-1G H0683-5G
TCIAH0683-1GEA 1675 SEK
TCIAH0683-1G TCIAH0683-5G
2-Hydroxy-5-methylisophthalaldehyde ≥98.0%
2-Hydroxy-5-methylisophthalaldehyde
Formel: C₉H₈O₃
MW: 164,1600 g/mol
MDL Number: MFCD00016596
CAS nummer: 7310-95-4
EINECS: 230-768-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c94cc9c8ee13f7275cdcc2d64d07fd261bbc5eb1&vid=c94cc9c8ee13f7275cdcc2d64d07fd261bbc5eb1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 129.0 to 133.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR