2-Bromoisophthalaldehyde ≥98.0%

Leverantör: TCI
Varning

B3568-1G B3568-5G
TCIAB3568-1GEA 550 SEK
TCIAB3568-1G TCIAB3568-5G
2-Bromoisophthalaldehyde ≥98.0%
2-Bromoisophthalaldehyde
Formel: C₈H₅BrO₂
MDL Number: MFCD08461034
CAS nummer: 79839-49-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8ac0ba078cb2654f34a2aa8817192804cda4a77&vid=e8ac0ba078cb2654f34a2aa8817192804cda4a77&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 137.0 to 143.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR