(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: (1-(Ethoxycarbonyl)cyclopropyl)triphenylphosphonium tetrafluoroborate
A18305.06 A18305.14
A18305.06EA 4400 SEK
A18305.06 A18305.14
(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate ≥98%
(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate
Purity: 98% CAS No.: 52186-89-7 Molecular Formula: C24H24BF4O2P Formula Weight: 462.23 Melting Point: 173-179° Beilstein Reference: 4650673
Formel: C₂₄H₂₄BF₄O₂P
MW: 462,23 g/mol
Smältpunkt: 173…179 °C
Storage Temperature: Omgivande
MDL Number: MFCD00051879
CAS nummer: 52186-89-7