3-(4-Fluorophenyl)-1H-pyrazol-5-amine 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 5-Amino-3-(4-fluorophenyl)-1H-pyrazole
H32830.03 H32830.06
H32830.03EA 1825 SEK
H32830.03 H32830.06
3-(4-Fluorophenyl)-1H-pyrazol-5-amine 97%
3-(4-Fluorophenyl)-1H-pyrazol-5-amine
Purity: 97% CAS No.: 72411-52-0 Molecular Formula: C9H8FN3 Formula Weight: 177.18 Melting Point: 122-125° MDL No.: MFCD01023677
Formel: C₉H₈FN₃
MW: 177,18 g/mol
Smältpunkt: 122…125 °C
Storage Temperature: Omgivande
MDL Number: MFCD01023677
CAS nummer: 72411-52-0