L(+)-Lyxose ≥98.0%

Leverantör: TCI EUROPE

L0153-1G L0153-5G
TCIAL0153-1GEA 1075 SEK
TCIAL0153-1G TCIAL0153-5G
L(+)-Lyxose ≥98.0%
L(+)-Lyxose
Formel: C₅H₁₀O₅
MW: 150,13 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00064111
CAS nummer: 1949-78-6
EINECS: 217-763-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67f17f18ece0305c2a94e49792d2a5cf029b8a9f&vid=67f17f18ece0305c2a94e49792d2a5cf029b8a9f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +13.0 to +15.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR