D-(-)-Lyxose ≥98.0%
Leverantör: TCI EUROPE
L0073-1G
L0073-25G
L0073-5G
TCIAL0073-1GEA
215
SEK
TCIAL0073-1G
TCIAL0073-25G
TCIAL0073-5G
D-(-)-Lyxose ≥98.0%
D-(-)-Lyxose
Formel:
C₅H₁₀O₅ MW: 150,13 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00064362 CAS nummer: 1114-34-7 EINECS: 214-212-8 Merck Index: 14,05641 |
Specifikation testresultat
Appearance | White to Almost white powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]20/D | -12.0 to -16.0 deg (C=1, H2O)[included NH3 aq. (2+3)] |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...