L(-)-Arabitol 97,0 GC_ANALYSMETOD

Leverantör: TCI

Synonyms: L-(-)-Arabinitol, (2S,4S)-Pentane-1,2,3,4,5-pentaol, L-(-)-Arabitol, L-Arabinitol

A0518-1G A0518-25G A0518-5G
TCIAA0518-1GEA 215 SEK
TCIAA0518-1G TCIAA0518-25G TCIAA0518-5G
L(-)-Arabitol 97,0 GC_ANALYSMETOD
L(-)-Arabitol
Formel: C₅H₁₂O₅
MW: 152.15 g/mol
MDL Number: MFCD00064290
CAS nummer: 7643-75-6
EINECS: 231-582-6
Merck Index: 14,00762
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c99bafc145e78d9e1c0387d323252ce95f0e1bc6&vid=c99bafc145e78d9e1c0387d323252ce95f0e1bc6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 101.0 to 104.0 deg-C
Specific rotation [a]20/D -10.0 to -14.0 deg (C=5, Sodium borate (8%))
Solubility in Water almost transparency

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR