L(-)-Arabitol 98

Leverantör: Thermo Fisher Scientific

Synonyms: L-(-)-Arabinitol, (2S,4S)-Pentane-1,2,3,4,5-pentaol, L-(-)-Arabitol, L-Arabinitol

A13103.06 A13103.14 A13103.22
A13103.06EA 1125 SEK
A13103.06 A13103.14 A13103.22
L(-)-Arabitol 98
L(-)-Arabitol

Purity: 98% CAS No.: 7643-75-6 Molecular Formula: C5H12O5 Formula Weight: 152.15 Melting Point: 98-103° Beilstein Reference: 1720521 Merck Reference: 14,762 Optical Rotation: -11° (c=5 in 8% borax soln) MDL No.: MFCD00064290

Formel: C₅H₁₂O₅
MW: 152.15 g/mol
Smältpunkt: 98…103 °C
Storage Temperature: Omgivande
MDL Number: MFCD00064290
CAS nummer: 7643-75-6
EINECS: 231-582-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR