D-(+)-Arabitol ≥98.0%

Leverantör: TCI EUROPE

Synonyms: D-Arabitol, (2R,4R)-Pentane-1,2,3,4,5-pentaol

A0516-1G A0516-25G
TCIAA0516-1GEA 215 SEK
TCIAA0516-1G TCIAA0516-25G
D-(+)-Arabitol ≥98.0%
D-(+)-Arabitol
Formel: C₅H₁₂O₅
MW: 152,15 g/mol
MDL Number: MFCD00004709
CAS nummer: 488-82-4
EINECS: 207-686-2
Merck Index: 14,00762
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d70a019b8baf2788fb9eb0cb2d42f2709de3782&vid=4d70a019b8baf2788fb9eb0cb2d42f2709de3782&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 102.0 to 105.0 deg-C
Specific rotation [a]20/D +10.0 to +14.0 deg (C=5, Sodium borate (8%))
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR