Arachidonic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: (5Z,8Z,11Z,14Z)-Icosa-5,8,11,14-tetraenoic acid

A0781-100MG A0781-500MG
TCIAA0781-100MGEA 990 SEK
TCIAA0781-100MG TCIAA0781-500MG
Arachidonic acid ≥98.0% (by GC, titration analysis)
Arachidonic acid
Formel: C₂₀H₃₂O₂
MW: 304,5 g/mol
Storage Temperature: Frys
MDL Number: MFCD00004417
CAS nummer: 506-32-1
EINECS: 208-033-4
Merck Index: 14,00765
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06ca9b0000530888f31860c9052067f86fb17fb1&vid=06ca9b0000530888f31860c9052067f86fb17fb1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR