Magnetic stirring bars, oval, PFTE
Leverantör: SciLabware
SWN520 SWN522 SWN524 SWN525 SWN526 SWN528
AZLOSWN520EA 430 SEK
AZLOSWN520 AZLOSWN522 AZLOSWN524 AZLOSWN525 AZLOSWN526 AZLOSWN528
Magnetic stirring bars, oval, PFTE
Omrörare Omrörarmagneter
PTFE covered, suitable for use in round bottomed flasks.
  • Very good chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)