2,7-Di-tert-butylfluorene ≥98.0%
Leverantör: TCI EUROPE
D4084-25G
D4084-5G
TCIAD4084-25GEA
695
SEK
TCIAD4084-25G
TCIAD4084-5G
2,7-Di-tert-butylfluorene ≥98.0%
2,7-Di-tert-butylfluorene
Formel:
C₂₁H₂₆ MW: 278,43 g/mol |
MDL Number:
MFCD03093998 CAS nummer: 58775-05-6 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 122.0 to 126.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...