Sodium bis(trimethylsilyl)amide 2 M i THF, AcroSeal®, ren
Leverantör: Thermo Scientific
Fara
Synonyms: HMDS sodium salt, N-Sodiohexamethyldisilazane, Sodium bis(trimethylsilyl)amide, HMDS-Na
277851000 277858000
277851000.EA 1775 SEK
277851000. 277858000.
Sodium bis(trimethylsilyl)amide 2 M i THF, AcroSeal®, ren
Sodium bis(trimethylsilyl)amide
Formel: C₆H₁₈NNaSi₂
MW: 183,38 g/mol
Flampunkt: –21 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009835
CAS nummer: 1070-89-9
EINECS: 213-983-8
UN: 2924
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear orange to brown Solution
Titration with HClO4 38 to 42 %