Isobutylmetakrylat ≥98.0% stabilised
Leverantör: TCI EUROPE
Varning
Synonyms: Methacrylic acid Isobutyl ester
Stabilization: med hydrokinon
M0132-25ML M0132-500ML
TCIAM0132-25MLEA 245 SEK
TCIAM0132-25ML TCIAM0132-500ML
Isobutylmetakrylat ≥98.0% stabilised
Isobutylmetakrylat
Formel: C₈H₁₄O₂
MW: 142,2 g/mol
Kokpunkt: 155°C
Flampunkt: 42°C
MDL Number: MFCD00008931
CAS nummer: 97-86-9
EINECS: 202-613-0
UN: 2283
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=749eb8a1de8d50513c8575f3a291612943413537&vid=749eb8a1de8d50513c8575f3a291612943413537&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless clear liquid
Purity (GC) min. 98.0 %