5-Bromo-2-ethoxypyridine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

B3673-5G B3673-1G
TCIAB3673-5GEA 990 SEK
TCIAB3673-5G TCIAB3673-1G
5-Bromo-2-ethoxypyridine ≥98.0% (by GC, titration analysis)
5-Bromo-2-ethoxypyridine
Formel: C₇H₈BrNO
MW: 202,05 g/mol
Kokpunkt: 107°C
MDL Number: MFCD00234311
CAS nummer: 55849-30-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4408a1c67ca301062f392dd9715d362ea980a514&vid=4408a1c67ca301062f392dd9715d362ea980a514&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 35.0 to 39.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR