2-Bromo-5-nitrobenzoic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

B3841-5G B3841-25G
TCIAB3841-5GEA 510 SEK
TCIAB3841-5G TCIAB3841-25G
2-Bromo-5-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
2-Bromo-5-nitrobenzoic acid
Formel: C₇H₄BrNO₄
Kokpunkt: 371 °C (1013 hPa)
Smältpunkt: 178…180 °C
MDL Number: MFCD00134558
CAS nummer: 943-14-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba1203e2068981072c22b537648c658ed2cb013a&vid=f7e10057ec9ecc43c8cb662a49345a1c5e449216&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 180.0 to 184.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR